Assay Details
Target Gene Details
Entrez Gene ID: | 23621 |
Gene Name: | beta-secretase 1 |
Gene Aliases: |
ASP2, BACE, HSPC104 |
Location: |
Chr.11:117285686-117316256 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACE1 | NM_001207048.1 | NP_001193977.1 | ||
| NM_001207049.1 | NP_001193978.1 | |||
| NM_012104.4 | NP_036236.1 | |||
| NM_138971.3 | NP_620427.1 | |||
| NM_138972.3 | NP_620428.1 | |||
| NM_138973.3 | NP_620429.1 | |||
| AB032975.2 | BAA86463.2 | |||
| AB050436.1 | BAB40931.1 | |||
| AB050437.1 | BAB40932.1 | |||
| AB050438.1 | BAB40933.1 | |||
| AB089958.1 | BAC81826.1 | |||
| AB177847.1 | BAD66825.1 | |||
| AF190725.1 | AAF04142.1 | |||
| AF200193.1 | AAF13715.1 | |||
| AF200343.1 | AAF17079.1 | |||
| AF201468.1 | AAF18982.1 | |||
| AF204943.1 | AAF26367.1 | |||
| AF338816.1 | AAK38374.1 | |||
| AF338817.1 | AAK38375.1 | |||
| AK075049.1 | ||||
| AK296015.1 | ||||
| AK296535.1 | ||||
| BC036084.1 | AAH36084.1 | |||
| BC065492.1 | AAH65492.1 | |||
| DC343475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038815 | Chr.11:117287202 - 117403925 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map