Assay Details
Target Gene Details
Entrez Gene ID: | 55176 |
Gene Name: | Sec61 translocon alpha 2 subunit |
Gene Aliases: |
- |
Location: |
Chr.10:12129641-12169958 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC61A2 | NM_001142627.2 | NP_001136099.1 | ||
| NM_001142628.1 | NP_001136100.1 | |||
| NM_018144.3 | NP_060614.2 | |||
| NR_024576.2 | ||||
| NR_024577.2 | ||||
| AF346603.1 | AAK29084.1 | |||
| AK022640.1 | BAB14148.1 | |||
| AK057532.1 | ||||
| AK292295.1 | ||||
| AK301841.1 | ||||
| BC026179.1 | AAH26179.1 | |||
| BP212860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv674n100 | Chr.10:12077012 - 12134998 on Build GRCh38 | Gain |
|
| nsv1035709 | Chr.10:12054410 - 12183299 on Build GRCh38 | Gain |
|
| dgv673n100 | Chr.10:12049505 - 12137652 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map