Assay Details
Target Gene Details
Entrez Gene ID: | 8891 |
Gene Name: | eukaryotic translation initiation factor 2B subunit gamma |
Gene Aliases: |
EIF-2B, EIF2Bgamma |
Location: |
Chr.1:44850522-44986722 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B3 | NM_001261418.1 | 11 | 1650 | NP_001248347.1 |
| NM_020365.4 | 12 | 1754 | NP_065098.1 | |
| XM_017002747.1 | XP_016858236.1 | |||
| AK022399.1 | BAB14027.1 | |||
| BC018728.1 | AAH18728.1 | |||
| BF879213.1 | 1 | 195 | ||
| BQ776018.1 | ||||
| W21485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546146 | Chr.1:44792469 - 44853080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map