Assay Details
Target Gene Details
Entrez Gene ID: | 1230 |
Gene Name: | C-C motif chemokine receptor 1 |
Gene Aliases: |
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1 |
Location: |
Chr.3:46201709-46208341 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR1 | NM_001295.2 | 2 | 906 | NP_001286.1 |
| BC051306.1 | 2 | 894 | AAH51306.2 | |
| BC064991.1 | 2 | 862 | AAH64991.1 | |
| BT019908.1 | 1 | 835 | AAV38711.1 | |
| BT019909.1 | 1 | 835 | AAV38712.1 | |
| BX337412.2 | 2 | 905 | ||
| D10925.1 | 1 | 857 | BAA01723.1 | |
| L09230.1 | 1 | 835 | AAA58408.1 | |
| L10918.1 | 2 | 897 | AAA36543.1 |
More Information
Set Membership: |
|

Genomic Map