Assay Details
Target Gene Details
Entrez Gene ID: | 51100 |
Gene Name: | SH3 domain containing GRB2 like endophilin B1 |
Gene Aliases: |
Bif-1, CGI-61, PPP1R70, dJ612B15.2 |
Location: |
Chr.1:86704570-86748184 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3GLB1 | NM_001206651.1 | 1 | 106 | NP_001193580.1 |
| NM_001206652.1 | 1 | 106 | NP_001193581.1 | |
| NM_001206653.1 | 1 | 106 | NP_001193582.1 | |
| NM_016009.4 | 1 | 106 | NP_057093.1 | |
| XM_006710672.2 | 1 | 106 | XP_006710735.1 | |
| XM_006710673.2 | 1 | 76 | XP_006710736.1 | |
| XM_011541538.1 | 1 | 106 | XP_011539840.1 | |
| XM_017001408.1 | 1 | 76 | XP_016856897.1 | |
| XM_017001409.1 | 1 | 106 | XP_016856898.1 | |
| XM_017001410.1 | 1 | 106 | XP_016856899.1 | |
| AB007960.2 | 1 | 71 | ||
| AK303710.1 | 1 | 59 | ||
| DB038227.1 | 1 | 106 | ||
| DB451777.1 | 1 | 102 |
More Information
Set Membership: |
|

Genomic Map