Assay Details
Target Gene Details
Entrez Gene ID: | 1654 |
Gene Name: | DEAD-box helicase 3, X-linked |
Gene Aliases: |
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102 |
Location: |
Chr.X:41333308-41364472 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX3X | NM_001193416.2 | NP_001180345.1 | ||
| NM_001193417.2 | NP_001180346.1 | |||
| NM_001356.4 | NP_001347.3 | |||
| NR_126093.1 | ||||
| XM_011543892.2 | XP_011542194.1 | |||
| XM_017029313.1 | XP_016884802.1 | |||
| XM_017029314.1 | XP_016884803.1 | |||
| AB208983.1 | BAD92220.1 | |||
| AB451220.1 | ||||
| AB451343.1 | ||||
| AF000982.1 | AAC51829.1 | |||
| AF000983.1 | AAC51830.1 | |||
| AF061337.1 | AAC34298.1 | |||
| AK291153.1 | ||||
| AK296906.1 | ||||
| AK297159.1 | ||||
| AK304661.1 | ||||
| AK304689.1 | ||||
| AK310891.1 | ||||
| BC011819.2 | AAH11819.1 | |||
| BM833870.1 | ||||
| U50553.1 | AAB95637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33720 | Chr.X:40891487 - 43157518 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map