Assay Details
Target Gene Details
Entrez Gene ID: | 10000 |
Gene Name: | AKT serine/threonine kinase 3 |
Gene Aliases: |
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2 |
Location: |
Chr.1:243488233-243851079 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKT3 | NM_181690.2 | NP_859029.1 | ||
| AY005799.1 | AAF91073.1 | |||
| BC121154.2 |
Target Gene Details
Entrez Gene ID: | 10806 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Location: |
Chr.1:243255328-243500092 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 24 - Intron 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833070 | Chr.1:243339809 - 243537501 on Build GRCh38 | Gain |
|
| nsv1013039 | Chr.1:243460621 - 243501269 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map