Assay Details
Target Gene Details
Entrez Gene ID: | 55687 |
Gene Name: | tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase |
Gene Aliases: |
LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1 |
Location: |
Chr.22:46335401-46357340 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMU | NM_001282782.1 | 10 | 1630 | NP_001269711.1 |
| NM_001282783.1 | 10 | 1571 | NP_001269712.1 | |
| NM_001282784.1 | 9 | 1488 | NP_001269713.1 | |
| NM_001282785.1 | 10 | 1654 | NP_001269714.1 | |
| NM_018006.4 | 11 | 1737 | NP_060476.2 | |
| NR_104240.1 | 11 | 1682 | ||
| NR_104241.1 | 10 | 1575 | ||
| XM_005261678.1 | 11 | 1365 | XP_005261735.1 | |
| XM_005261681.1 | 10 | 1282 | XP_005261738.1 | |
| XM_011530271.2 | 11 | 1500 | XP_011528573.1 | |
| XM_011530272.1 | 10 | 1323 | XP_011528574.1 | |
| XM_011530273.1 | 9 | 1240 | XP_011528575.1 | |
| XM_011530274.2 | 10 | 1337 | XP_011528576.1 | |
| XM_011530275.1 | 11 | 1321 | XP_011528577.1 | |
| AB178028.1 | 11 | 1424 | BAD66875.1 | |
| AK000212.1 | 2 | 1635 | ||
| AK001002.1 | 11 | 1369 | BAA91462.1 | |
| AK055295.1 | 2 | 2709 | ||
| AK093743.1 | 3 | 2427 | ||
| AK290712.1 | 11 | 1417 | ||
| AY062123.2 | 11 | 1713 | AAL38183.1 | |
| BC027991.1 | 10 | 1229 | ||
| BM755897.1 | 1 | 267 | ||
| CR456445.1 | 10 | 1310 | CAG30331.1 | |
| DQ309998.1 | 10 | 1551 | ABC46690.1 | |
| DQ309999.1 | 10 | 1606 | ABC46691.1 | |
| DQ310000.1 | 9 | 1464 | ABC46692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955177 | Chr.22:46350404 - 46389303 on Build GRCh38 | Deletion |
|
| nsv829293 | Chr.22:46241054 - 46741639 on Build GRCh38 | Gain |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
| nsv834225 | Chr.22:46323584 - 46510799 on Build GRCh38 | Loss |
|
| nsv471214 | Chr.22:46278315 - 46367774 on Build GRCh38 | Loss |
|
| nsv524100 | Chr.22:46356410 - 46357282 on Build GRCh38 | Loss |
|
| nsv1066865 | Chr.22:46352413 - 46490139 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map