Assay Details
Target Gene Details
Entrez Gene ID: | 9311 |
Gene Name: | acid sensing ion channel subunit 3 |
Gene Aliases: |
ACCN3, DRASIC, SLNAC1, TNaC1 |
Location: |
Chr.7:151048292-151052756 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC3 | NM_004769.3 | NP_004760.1 | ||
| NM_020321.3 | NP_064717.1 | |||
| NM_020322.3 | NP_064718.1 | |||
| NR_046401.1 | ||||
| AB010575.1 | BAA25897.1 | |||
| AB209421.1 | BAD92658.1 | |||
| AF057711.1 | AAC62935.1 | |||
| AF095897.1 | AAC64188.1 | |||
| AF195024.1 | AAF19817.1 | |||
| AF195025.1 | AAF19818.1 | |||
| AK128129.1 | 22 | 4107 | ||
| AK313926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
| nsv526410 | Chr.7:151017725 - 151078219 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| dgv6756n100 | Chr.7:151033912 - 151093430 on Build GRCh38 | Gain |
|
| nsv524928 | Chr.7:151034611 - 151069712 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map