Assay Details
Target Gene Details
Entrez Gene ID: | 4284 |
Gene Name: | major intrinsic protein of lens fiber |
Gene Aliases: |
AQP0, CTRCT15, LIM1, MIP26, MP26 |
Location: |
Chr.12:56449502-56456553 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIP | NM_012064.3 | 4 | 721 | NP_036196.1 |
| XM_011538354.1 | 6 | 538 | XP_011536656.1 | |
| XM_017019306.1 | 4 | 462 | XP_016874795.1 | |
| BC074913.2 | 4 | 712 | AAH74913.1 | |
| BC117474.1 | 4 | 721 | ||
| KT966395.1 | 4 | 670 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map