Assay Details
Target Gene Details
Entrez Gene ID: | 23541 |
Gene Name: | SEC14 like lipid binding 2 |
Gene Aliases: |
C22orf6, SPF, TAP, TAP1 |
Location: |
Chr.22:30396941-30425303 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L2 | NM_001204204.2 | NP_001191133.1 | ||
| NM_001291932.1 | 4 | 430 | NP_001278861.1 | |
| NM_012429.4 | 5 | 506 | NP_036561.1 | |
| NM_033382.2 | 5 | 506 | NP_203740.1 | |
| AB033012.2 | 5 | 405 | ||
| AK091402.1 | 3 | 293 | ||
| AK223587.1 | 5 | 466 | BAD97307.1 | |
| AK296540.1 | 4 | 320 | ||
| AK303751.1 | ||||
| AK314390.1 | 5 | 439 | ||
| AL096881.1 | 5 | 503 | CAB51405.1 | |
| AY927438.1 | 4 | 235 | ||
| AY927439.1 | 4 | 235 | ||
| BC058915.1 | 5 | 410 | AAH58915.1 | |
| CR456571.1 | 5 | 403 | CAG30457.1 | |
| DC375707.1 | 4 | 430 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map