Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMS2 | NM_002898.3 | NP_002889.1 | ||
| XM_005269059.4 | XP_005269116.1 | |||
| XM_005269060.4 | XP_005269117.1 | |||
| XM_005269061.3 | XP_005269118.1 | |||
| XM_005269066.4 | XP_005269123.1 | |||
| XM_006719541.3 | XP_006719604.1 | |||
| XM_006719542.3 | XP_006719605.1 | |||
| XM_006719543.3 | XP_006719606.1 | |||
| XM_006719544.3 | XP_006719607.1 | |||
| XM_011538638.2 | XP_011536940.1 | |||
| XM_011538639.2 | XP_011536941.1 | |||
| XM_011538640.2 | XP_011536942.1 | |||
| AK293315.1 | ||||
| AK299580.1 | ||||
| AK304540.1 | ||||
| AK308885.1 | ||||
| BC027863.1 | AAH27863.1 | |||
| BC072679.1 | AAH72679.1 | |||
| D28483.1 | BAA05842.1 | |||
| DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv1052713 | Chr.12:56454252 - 56523818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map