Assay Details
Target Gene Details
Entrez Gene ID: | 283165 |
Gene Name: | KIRREL3 antisense RNA 3 |
Gene Aliases: |
NCRNA00288, PRR10 |
Location: |
Chr.11:127002910-127006058 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIRREL3-AS3 | NR_040078.1 | |||
| AK097543.1 | BAC05095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv539 | Chr.11:126984853 - 127009528 on Build GRCh38 | Insertion |
|
| esv2761719 | Chr.11:126950790 - 127014459 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map