Assay Details
Target Gene Details
Entrez Gene ID: | 28976 |
Gene Name: | acyl-CoA dehydrogenase family member 9 |
Gene Aliases: |
NPD002 |
Location: |
Chr.3:128879490-128916075 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAD9 | NM_014049.4 | 1 | 272 | NP_054768.2 |
| NR_033426.1 | 1 | 272 | ||
| XM_017006264.1 | 1 | 166 | XP_016861753.1 | |
| AB209427.1 | 1 | 119 | BAD92664.1 | |
| AF327351.1 | 1 | 102 | AAL56011.1 | |
| AK022568.1 | 1 | 116 | BAB14104.1 | |
| AK024012.1 | 1 | 116 | BAB14775.1 | |
| AK130311.1 | 1 | 143 | ||
| AK294597.1 | 1 | 103 | ||
| AK297952.1 | 1 | 140 | ||
| AK310644.1 | 1 | 139 | ||
| BC007970.2 | 1 | 151 | AAH07970.1 | |
| BC013354.2 | 1 | 126 | AAH13354.1 | |
| DA630022.1 | 1 | 272 |
More Information
Set Membership: |
|

Genomic Map