Assay Details
Target Gene Details
Entrez Gene ID: | 23649 |
Gene Name: | DNA polymerase alpha 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.11:65261852-65306269 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLA2 | NM_002689.3 | NP_002680.2 | ||
| XM_011544877.2 | XP_011543179.1 | |||
| XM_011544878.2 | XP_011543180.1 | |||
| XM_011544879.1 | XP_011543181.1 | |||
| XM_011544881.2 | XP_011543183.1 | |||
| XM_017017452.1 | XP_016872941.1 | |||
| AK025315.1 | ||||
| AK297842.1 | ||||
| AK307831.1 | ||||
| AK309284.1 | ||||
| BC001347.2 | AAH01347.1 | |||
| BC002990.1 | AAH02990.1 | |||
| CR456737.1 | CAG33018.1 | |||
| L24559.1 | AAA16459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832190 | Chr.11:65210673 - 65360142 on Build GRCh38 | Loss |
|
| esv2422285 | Chr.11:65245823 - 65487069 on Build GRCh38 | Deletion |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map