Assay Details
Target Gene Details
Entrez Gene ID: | 7799 |
Gene Name: | PR domain 2 |
Gene Aliases: |
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2 |
Location: |
Chr.1:13698875-13825079 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM2 | NM_001135610.1 | 5 | 420 | NP_001129082.1 |
| NM_012231.4 | 5 | 1150 | NP_036363.2 | |
| NM_015866.4 | 5 | 1150 | NP_056950.2 | |
| XM_017002253.1 | 4 | 372 | XP_016857742.1 | |
| XM_017002254.1 | 4 | 372 | XP_016857743.1 | |
| XM_017002255.1 | 4 | 382 | XP_016857744.1 | |
| XM_017002256.1 | 5 | 451 | XP_016857745.1 | |
| XM_017002257.1 | 4 | 380 | XP_016857746.1 | |
| XM_017002258.1 | 2 | 82 | XP_016857747.1 | |
| XM_017002264.1 | 4 | 1711 | XP_016857753.1 | |
| U17838.1 | 5 | 1149 | AAC50820.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545567 | Chr.1:13558784 - 14026576 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map