Assay Details
Target Gene Details
Entrez Gene ID: | 374659 |
Gene Name: | HD domain containing 3 |
Gene Aliases: |
(ppGpp)ase, MESH1 |
Location: |
Chr.15:90930918-90932569 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDDC3 | NM_001286451.1 | NP_001273380.1 | ||
| NM_198527.3 | 4 | 594 | NP_940929.1 | |
| NR_104447.1 | ||||
| AK057584.1 | 1 | 1162 | ||
| BC033794.1 | 4 | 594 | AAH33794.1 | |
| BQ777217.1 | ||||
| BU633661.1 | 1 | 493 | ||
| CA450487.1 | ||||
| CV573934.1 |
Target Gene Details
Entrez Gene ID: | 55898 |
Gene Name: | unc-45 myosin chaperone A |
Gene Aliases: |
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A |
Location: |
Chr.15:90929980-90954093 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC45A | NM_001039675.1 | NP_001034764.1 | ||
| XM_011521779.2 | XP_011520081.1 | |||
| AK125721.1 | ||||
| BC045635.1 | AAH45635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833090 | Chr.15:90911195 - 91098519 on Build GRCh38 | Loss |
|
| nsv515661 | Chr.15:90866779 - 90981967 on Build GRCh38 | Loss |
|
| nsv833089 | Chr.15:90859910 - 90962961 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map