Assay Details
Target Gene Details
Entrez Gene ID: | 272 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.11:10450321-10507579 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMPD3 | NM_000480.2 | 3 | 745 | NP_000471.1 |
| NM_001025389.1 | 3 | 708 | NP_001020560.1 | |
| NM_001025390.1 | 3 | 847 | NP_001020561.1 | |
| NM_001172430.1 | 3 | 672 | NP_001165901.1 | |
| NM_001172431.1 | NP_001165902.1 | |||
| AK289998.1 | 3 | 479 | ||
| AK294444.1 | ||||
| AK295046.1 | ||||
| AK299113.1 | 3 | 420 | ||
| AK301507.1 | 3 | 595 | ||
| AK302970.1 | 3 | 663 | ||
| AK308181.1 | 3 | 515 | ||
| AK311436.1 | 4 | 552 | ||
| AK311607.1 | 3 | 768 | ||
| AK311711.1 | 3 | 732 | ||
| AK313039.1 | 3 | 479 | ||
| BC126118.1 | 3 | 740 | ||
| D12775.1 | 3 | 564 | BAA02240.1 | |
| M84720.1 | 3 | 579 | AAA58365.1 | |
| M84721.1 | 3 | 507 | AAA58366.1 | |
| M84722.1 | 3 | 631 | AAA58367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526453 | Chr.11:10477556 - 10480875 on Build GRCh38 | Loss |
|
| esv3625358 | Chr.11:10435119 - 10512909 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map