Assay Details
Target Gene Details
Entrez Gene ID: | 387837 |
Gene Name: | C-type lectin domain family 12 member B |
Gene Aliases: |
UNQ5782 |
Location: |
Chr.12:10006138-10018800 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC12B | NM_001129998.2 | 1 | 35 | NP_001123470.1 |
| NM_001319241.1 | 1 | 35 | NP_001306170.1 | |
| NM_001319242.1 | 1 | 35 | NP_001306171.1 | |
| NM_205852.3 | 1 | 35 | NP_995324.2 | |
| NR_135049.1 | 1 | 35 | ||
| XM_011520658.2 | XP_011518960.1 | |||
| XM_011520663.2 | 2 | 126 | XP_011518965.1 | |
| XM_017019295.1 | 2 | 115 | XP_016874784.1 | |
| AK128243.1 | BAC87350.1 | |||
| AY358810.1 | 1 | 35 | AAQ89170.1 | |
| BC136805.1 | ||||
| BC136806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1374n100 | Chr.12:9968689 - 10026775 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map