Assay Details
Target Gene Details
Entrez Gene ID: | 10283 |
Gene Name: | CWC27 spliceosome associated protein homolog |
Gene Aliases: |
NY-CO-10, SDCCAG-10, SDCCAG10 |
Location: |
Chr.5:64768918-65018763 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWC27 | NM_001297644.1 | NP_001284573.1 | ||
| NM_001297645.1 | NP_001284574.1 | |||
| NM_001318000.1 | NP_001304929.1 | |||
| NM_005869.3 | NP_005860.2 | |||
| XM_011543095.2 | XP_011541397.2 | |||
| XM_011543096.1 | XP_011541398.1 | |||
| XM_017008943.1 | XP_016864432.1 | |||
| AK225105.1 | ||||
| AK225161.1 | ||||
| AK308223.1 | ||||
| AK312954.1 | ||||
| AY358569.1 | AAQ88932.1 | |||
| BX647971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1031566 | Chr.5:64737106 - 64787266 on Build GRCh38 | Gain |
|
| nsv598390 | Chr.5:64554330 - 64982002 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map