Assay Details
Target Gene Details
Entrez Gene ID: | 23457 |
Gene Name: | ATP binding cassette subfamily B member 9 |
Gene Aliases: |
EST122234, TAPL |
Location: |
Chr.12:122918708-122975160 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB9 | XM_011538095.2 | 1 | 106 | XP_011536397.1 |
| AB040953.2 | ||||
| BC064384.1 | 1 | 151 | AAH64384.1 |
Target Gene Details
Entrez Gene ID: | 79676 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:122974703-122980041 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGFOD2 | NM_001304834.1 | 1 | 353 | NP_001291763.1 |
| NM_001304835.1 | 1 | 353 | NP_001291764.1 | |
| NM_001304836.1 | 1 | 353 | NP_001291765.1 | |
| NM_001304837.1 | 1 | 353 | NP_001291766.1 | |
| NM_001304838.1 | 1 | 353 | NP_001291767.1 | |
| NM_024623.2 | 1 | 353 | NP_078899.1 | |
| AK023553.1 | 1 | 249 | BAB14610.1 | |
| AK095481.1 | 1 | 261 | ||
| BC098119.1 | 1 | 85 | AAH98119.1 | |
| BC098293.1 | 1 | 85 | AAH98293.1 | |
| BC101940.1 | 1 | 85 | AAI01941.1 | |
| BC105637.1 | 1 | 85 | AAI05638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv455732 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map