Assay Details
Target Gene Details
Entrez Gene ID: | 8521 |
Gene Name: | glial cells missing homolog 1 |
Gene Aliases: |
GCMA, hGCMa |
Location: |
Chr.6:53125644-53262803 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCM1 | NM_003643.3 | 6 | 1081 | NP_003634.2 |
| XM_017011390.1 | 10 | 2463 | XP_016866879.1 | |
| AB026493.1 | 5 | 952 | BAA77250.2 | |
| AB041714.1 | 5 | 872 | BAA94757.1 | |
| BC096288.1 | 6 | 1039 | AAH96288.1 | |
| D88613.1 | 6 | 1084 | BAA13651.1 |
More Information
Set Membership: |
|

Genomic Map