Assay Details
Target Gene Details
Entrez Gene ID: | 9470 |
Gene Name: | eukaryotic translation initiation factor 4E family member 2 |
Gene Aliases: |
4E-LP, 4EHP, EIF4EL3, IF4e |
Location: |
Chr.2:232550587-232583645 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E2 | NM_001276336.1 | NP_001263265.1 | ||
| NM_001276337.1 | NP_001263266.1 | |||
| NM_001282958.1 | NP_001269887.1 | |||
| NM_004846.3 | NP_004837.1 | |||
| XM_005246975.3 | XP_005247032.1 | |||
| XM_011512206.2 | XP_011510508.1 | |||
| XM_017005377.1 | XP_016860866.1 | |||
| AB209519.1 | BAD92756.1 | |||
| AF038957.1 | AAC39871.1 | |||
| AF047695.1 | AAC18565.1 | |||
| AF068117.1 | AAC19374.1 | |||
| AK022518.1 | ||||
| AK303802.1 | ||||
| BC005392.1 | AAH05392.1 | |||
| BC005874.2 | AAH05874.1 | |||
| BC021226.2 | AAH21226.1 | |||
| BC021690.1 | AAH21690.1 | |||
| BG913475.1 | ||||
| BM018173.1 | ||||
| CD684451.1 | ||||
| DA408433.1 | ||||
| W80980.1 |
More Information
Set Membership: |
|

Genomic Map