Assay Details
Target Gene Details
Entrez Gene ID: | 64375 |
Gene Name: | IKAROS family zinc finger 4 |
Gene Aliases: |
EOS, ZNFN1A4 |
Location: |
Chr.12:56007647-56038435 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Gene Details
Entrez Gene ID: | 105369781 |
Gene Name: | uncharacterized LOC105369781 |
Gene Aliases: |
- |
Location: |
Chr.12:56010092-56025091 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369781 | NR_135023.1 | |||
| HY183314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
| nsv832427 | Chr.12:55998761 - 56118990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map