Assay Details
Target Gene Details
Entrez Gene ID: | 26206 |
Gene Name: | sperm associated antigen 8 |
Gene Aliases: |
BS-84, CILD28, CT142, HSD-1, SMP1, SPAG3, hSMP-1 |
Location: |
Chr.9:35807785-35812262 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG8 | NM_001039592.1 | 1 | 22 | NP_001034681.1 |
| NM_172312.1 | 1 | 22 | NP_758516.1 | |
| AK302313.1 | ||||
| BI757195.1 | ||||
| S83157.1 | 1 | 23 | AAB46833.1 | |
| U12978.2 | 1 | 23 | AAA21129.2 |
More Information
Set Membership: |
|

Genomic Map