Assay Details
Target Gene Details
Entrez Gene ID: | 84867 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 5 |
Gene Aliases: |
PTPSTEP, STEP, STEP61 |
Location: |
Chr.11:18727928-18792721 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN5 | NM_001039970.1 | 15 | 2303 | NP_001035059.1 |
| NM_001278236.1 | 15 | 3137 | NP_001265165.1 | |
| NM_001278238.1 | 14 | 2266 | NP_001265167.1 | |
| NM_001278239.1 | 14 | 2170 | NP_001265168.1 | |
| NM_006906.1 | 15 | 2399 | NP_008837.1 | |
| NM_032781.3 | 15 | 2354 | NP_116170.3 | |
| XM_011520411.2 | 15 | 2385 | XP_011518713.1 | |
| XM_017018434.1 | 14 | 2177 | XP_016873923.1 | |
| XM_017018435.1 | 14 | 2147 | XP_016873924.1 | |
| XM_017018436.1 | 13 | 2083 | XP_016873925.1 | |
| XM_017018437.1 | 14 | 2101 | XP_016873926.1 | |
| XM_017018438.1 | 12 | 1862 | XP_016873927.1 | |
| XM_017018439.1 | 13 | 1988 | XP_016873928.1 | |
| XM_017018440.1 | 14 | 2224 | XP_016873929.1 | |
| AK027333.1 | 1 | 340 | ||
| AK055450.1 | 3 | 1565 | ||
| AK090923.1 | 15 | 2263 | BAC03548.1 | |
| AK127312.1 | 15 | 3137 | ||
| AK294374.1 | 14 | 2006 | ||
| AK294673.1 | 14 | 1967 | ||
| AK295604.1 | 14 | 2127 | ||
| AL832541.1 | 15 | 2398 | CAD38632.2 | |
| BC039897.1 | 14 | 2062 | AAH39897.1 | |
| BC046435.1 | 5 | 878 | AAH46435.1 | |
| BC064807.1 | 15 | 2163 | AAH64807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047371 | Chr.11:18709926 - 18741490 on Build GRCh38 | Gain |
|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss |
|
| nsv525452 | Chr.11:18714400 - 18780956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map