Assay Details
Target Gene Details
Entrez Gene ID: | 5146 |
Gene Name: | phosphodiesterase 6C |
Gene Aliases: |
ACHM5, COD4, PDEA2 |
Location: |
Chr.10:93612588-93665673 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6C | NM_006204.3 | 1 | 265 | NP_006195.3 |
| U31973.1 | 1 | 265 | AAA96392.1 |
More Information
Set Membership: |
|

Genomic Map