Assay Details
Target Gene Details
Entrez Gene ID: | 113510 |
Gene Name: | helicase, POLQ-like |
Gene Aliases: |
HEL308 |
Location: |
Chr.4:83407343-83455883 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HELQ | NM_001297755.1 | NP_001284684.1 | ||
| NM_001297756.1 | NP_001284685.1 | |||
| NM_001297757.1 | NP_001284686.1 | |||
| NM_133636.3 | NP_598375.2 | |||
| XM_005262711.1 | XP_005262768.1 | |||
| XM_005262713.2 | XP_005262770.1 | |||
| XM_017007679.1 | XP_016863168.1 | |||
| XM_017007680.1 | XP_016863169.1 | |||
| XM_017007682.1 | XP_016863171.1 | |||
| XM_017007683.1 | XP_016863172.1 | |||
| AF436845.1 | AAL85274.1 | |||
| AK308395.1 | ||||
| AK312725.1 | ||||
| AL512702.1 | CAH56359.1 | |||
| CK905810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893971 | Chr.4:83096792 - 83493129 on Build GRCh38 | Gain |
|
| nsv829992 | Chr.4:83244588 - 83458578 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map