Assay Details
Target Gene Details
Entrez Gene ID: | 60681 |
Gene Name: | FK506 binding protein 10 |
Gene Aliases: |
BRKS1, FKBP65, OI11, OI6, PPIASE, hFKBP65 |
Location: |
Chr.17:41812262-41823217 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP10 | NM_021939.3 | 3 | 761 | NP_068758.3 |
| XM_011525099.2 | 3 | 1209 | XP_011523401.1 | |
| XM_011525100.2 | 2 | 336 | XP_011523402.1 | |
| AB045981.1 | 3 | 522 | BAB20974.1 | |
| AF337909.1 | 3 | 436 | AAM15770.1 | |
| AK025694.1 | 2 | 193 | BAB15220.1 | |
| AK092042.1 | 2 | 336 | ||
| AK092708.1 | 3 | 761 | BAC03954.1 | |
| AK172811.1 | 3 | 467 | BAD18781.1 | |
| AK296107.1 | ||||
| AL832985.1 | 3 | 503 | CAH56332.1 | |
| BC016467.1 | 3 | 496 | AAH16467.1 | |
| BX537452.1 | 3 | 516 | CAD97695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060502 | Chr.17:41767306 - 41821751 on Build GRCh38 | Gain |
|
| dgv3174n100 | Chr.17:41778258 - 41843183 on Build GRCh38 | Gain |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map