Assay Details
Target Gene Details
Entrez Gene ID: | 57505 |
Gene Name: | alanyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
AARSL, COXPD8, LKENP, MT-ALARS, MTALARS |
Location: |
Chr.6:44297850-44313358 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AARS2 | NM_020745.3 | NP_065796.1 | ||
| XM_005249245.3 | XP_005249302.1 | |||
| XM_011514764.2 | XP_011513066.1 | |||
| XM_017011112.1 | XP_016866601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv10641n54 | Chr.6:44224183 - 44314579 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79454687] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map