Assay Details
Target Gene Details
Entrez Gene ID: | 339906 |
Gene Name: | protease, serine 42 |
Gene Aliases: |
TESSP2 |
Location: |
Chr.3:46830404-46834095 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS42 | NM_182702.1 | NP_874361.1 | ||
| AJ544583.1 | CAD67566.1 | |||
| AK092703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv102e203 | Chr.3:46747685 - 46838512 on Build GRCh38 | Loss |
|
| nsv508915 | Chr.3:46814349 - 46913473 on Build GRCh38 | Insertion |
|
| esv2759147 | Chr.3:46638962 - 46930999 on Build GRCh38 | Loss |
|
| dgv4732n100 | Chr.3:46750043 - 46838650 on Build GRCh38 | Loss |
|
| nsv590204 | Chr.3:46811189 - 46860164 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map