Assay Details
Target Gene Details
Entrez Gene ID: | 56171 |
Gene Name: | dynein axonemal heavy chain 7 |
Gene Aliases: |
- |
Location: |
Chr.2:195737604-196075188 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 61 - Exon 62 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAH7 | NM_018897.2 | NP_061720.2 | ||
| XM_011511487.2 | XP_011509789.1 | |||
| XM_011511488.2 | XP_011509790.1 | |||
| XM_011511489.2 | XP_011509791.1 | |||
| XM_011511490.2 | XP_011509792.1 | |||
| XM_011511497.2 | XP_011509799.1 | |||
| XM_017004504.1 | XP_016859993.1 | |||
| AB023161.2 | ||||
| AF327442.1 | AAL37427.1 | |||
| BC029567.1 | AAH29567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834501 | Chr.2:195608503 - 195778199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map