Assay Details
Target Gene Details
Entrez Gene ID: | 10455 |
Gene Name: | enoyl-CoA delta isomerase 2 |
Gene Aliases: |
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3 |
Location: |
Chr.6:4115693-4135597 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECI2 | NM_001166010.1 | NP_001159482.1 | ||
| NM_006117.2 | NP_006108.2 | |||
| NM_206836.2 | NP_996667.2 | |||
| NR_028588.1 | ||||
| XM_006714957.1 | XP_006715020.1 | |||
| XM_017010170.1 | XP_016865659.1 | |||
| AB209917.1 | BAD93154.1 | |||
| AF069301.1 | AAC19317.1 | |||
| AF153612.1 | AAD34173.1 | |||
| AF244138.1 | AAF66247.1 | |||
| AF257175.1 | AAF68974.1 | |||
| AK057175.1 | ||||
| AK075108.1 | ||||
| AK222617.1 | BAD96337.1 | |||
| AK223003.1 | BAD96723.1 | |||
| AK297049.1 | ||||
| AL136642.1 | CAB66577.1 | |||
| AU100345.1 | ||||
| AY297537.1 | ||||
| BC002668.2 | AAH02668.3 | |||
| BC016781.1 | AAH16781.1 | |||
| BC017474.1 | AAH17474.1 | |||
| BC033841.1 | AAH33841.3 | |||
| BC034702.1 | AAH34702.1 | |||
| CR456768.1 | CAG33049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422457 | Chr.6:4100099 - 4377162 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map