Assay Details
Target Gene Details
Entrez Gene ID: | 101926934 |
Gene Name: | uncharacterized LOC101926934 |
Gene Aliases: |
- |
Location: |
Chr.6:46670444-46688165 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926934 | NR_134642.1 | |||
| NR_134643.1 | ||||
| NR_134644.1 | ||||
| AA725776.1 | ||||
| AI638322.1 | ||||
| AI698692.1 | ||||
| BX114358.1 |
Target Gene Details
Entrez Gene ID: | 9481 |
Gene Name: | solute carrier family 25 member 27 |
Gene Aliases: |
UCP4 |
Location: |
Chr.6:46652915-46678190 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A27 | NM_001204051.1 | NP_001190980.1 | ||
| NM_001204052.1 | NP_001190981.1 | |||
| NM_004277.4 | NP_004268.3 | |||
| XM_005249483.2 | XP_005249540.1 | |||
| AK090871.1 |
More Information
Set Membership: |
|

Genomic Map