Assay Details
Target Gene Details
Entrez Gene ID: | 8668 |
Gene Name: | eukaryotic translation initiation factor 3 subunit I |
Gene Aliases: |
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36 |
Location: |
Chr.1:32221584-32231604 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3I | NM_003757.3 | NP_003748.1 | ||
| XM_017002671.1 | XP_016858160.1 | |||
| AK130633.1 | ||||
| AK222483.1 | BAD96203.1 | |||
| AK225024.1 | ||||
| AK289356.1 | ||||
| AK289882.1 | ||||
| BC000413.2 | AAH00413.1 | |||
| BC003140.1 | AAH03140.1 | |||
| BM761611.1 | ||||
| BT019597.1 | AAV38404.1 | |||
| U36764.1 | AAC50224.1 | |||
| U39067.1 | AAC97144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map