Assay Details
Target Gene Details
Entrez Gene ID: | 148581 |
Gene Name: | ubiquitin conjugating enzyme E2 U (putative) |
Gene Aliases: |
- |
Location: |
Chr.1:64202802-64267381 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2U | NM_152489.1 | 1 | 205 | NP_689702.1 |
| XM_005270498.4 | 1 | 999 | XP_005270555.1 | |
| XM_011540764.2 | 2 | 940 | XP_011539066.1 | |
| BC029895.1 | 1 | 205 | AAH29895.1 |
More Information
Set Membership: |
|

Genomic Map