Assay Details
Target Gene Details
Entrez Gene ID: | 5624 |
Gene Name: | protein C, inactivator of coagulation factors Va and VIIIa |
Gene Aliases: |
APC, PC, PROC1, THPH3, THPH4 |
Location: |
Chr.2:127418221-127429246 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROC | NM_000312.3 | 9 | 1499 | NP_000303.1 |
| XM_005263717.4 | 9 | 1495 | XP_005263774.1 | |
| XM_017004505.1 | 9 | 1663 | XP_016859994.1 | |
| XM_017004506.1 | 7 | 1524 | XP_016859995.1 | |
| AK298280.1 | 9 | 1559 | ||
| AK298449.1 | 8 | 1593 | ||
| AK298454.1 | 8 | 1578 | ||
| AK303773.1 | 8 | 1556 | ||
| BC034377.1 | 9 | 1460 | AAH34377.1 | |
| BM975043.1 | 1 | 299 | ||
| K02059.1 | 5 | 1091 | AAA60164.1 | |
| S50739.1 | 1 | 163 | AAB24150.1 | |
| X02750.1 | 9 | 1502 | CAA26528.1 |
More Information
Set Membership: |
|

Genomic Map