Assay Details
Target Gene Details
Entrez Gene ID: | 10075 |
Gene Name: | HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase |
Gene Aliases: |
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1 |
Location: |
Chr.X:53532096-53686729 on Build GRCh38 |
Assay Gene Location: | Within Exon 79 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| esv3576879 | Chr.X:53529972 - 53593214 on Build GRCh38 | Gain |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map