Assay Details
Target Gene Details
Entrez Gene ID: | 55293 |
Gene Name: | UEV and lactate/malate dehyrogenase domains |
Gene Aliases: |
ATTP, UEV3 |
Location: |
Chr.11:18529609-18588837 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UEVLD | NM_001040697.2 | 8 | 933 | NP_001035787.1 |
| NM_001261382.1 | 7 | 867 | NP_001248311.1 | |
| NM_001261383.1 | 7 | 867 | NP_001248312.1 | |
| NM_001261384.1 | 6 | 684 | NP_001248313.1 | |
| NM_001261385.1 | 8 | 890 | NP_001248314.1 | |
| NM_001261386.1 | NP_001248315.1 | |||
| NM_018314.4 | 8 | 933 | NP_060784.3 | |
| XM_006718255.2 | 7 | 762 | XP_006718318.1 | |
| XM_011520201.2 | 4 | 2764 | XP_011518503.1 | |
| XM_017017985.1 | 7 | 860 | XP_016873474.1 | |
| XM_017017986.1 | 7 | 740 | XP_016873475.1 | |
| AF503350.1 | 8 | 893 | AAN32950.1 | |
| AK001930.1 | 8 | 890 | BAA91985.1 | |
| AK295501.1 | 6 | 612 | ||
| AK296835.1 | 7 | 827 | ||
| AK301537.1 | 7 | 809 | ||
| AK314521.1 | 8 | 933 | ||
| AK316191.1 | 7 | 740 | ||
| BC064566.1 | 7 | 807 | AAH64566.1 | |
| BU633352.1 | ||||
| CD109744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map