Assay Details
Target Gene Details
Entrez Gene ID: | 56945 |
Gene Name: | mitochondrial ribosomal protein S22 |
Gene Aliases: |
C3orf5, COXPD5, GIBT, GK002, MRP-S22, RPMS22 |
Location: |
Chr.3:139343994-139357129 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS22 | NM_020191.2 | 1 | 122 | NP_064576.1 |
| XM_005247640.2 | 1 | 121 | XP_005247697.1 | |
| XM_006713703.3 | 1 | 147 | XP_006713766.1 | |
| AB208981.1 | BAD92218.1 | |||
| AF063603.1 | 1 | 119 | AAG43162.1 | |
| AF226045.1 | 1 | 122 | AAF86945.1 | |
| AF321613.1 | 3 | 546 | AAK01406.1 | |
| AK075436.1 | 1 | 114 | BAC11619.1 | |
| AK292852.1 | 1 | 121 | ||
| BC009296.2 | 1 | 117 | AAH09296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005158 | Chr.3:139282894 - 139354169 on Build GRCh38 | Gain |
|
| nsv4034 | Chr.3:139316134 - 139361291 on Build GRCh38 | Deletion |
|
| nsv4035 | Chr.3:139321680 - 139355530 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map