Assay Details
Target Gene Details
Entrez Gene ID: | 56172 |
Gene Name: | ANKH inorganic pyrophosphate transport regulator |
Gene Aliases: |
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK |
Location: |
Chr.5:14704800-14871778 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKH | NM_054027.4 | 12 | 3526 | NP_473368.1 |
| XM_017009644.1 | 12 | 3126 | XP_016865133.1 | |
| AB046801.1 | 12 | 3356 | ||
| AK001799.1 |
Target Gene Details
Entrez Gene ID: | 90268 |
Gene Name: | OTU deubiquitinase with linear linkage specificity |
Gene Aliases: |
FAM105B, GUM |
Location: |
Chr.5:14660794-14716552 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTULIN | XM_011514151.2 | XP_011512453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026914 | Chr.5:14237673 - 14751479 on Build GRCh38 | Gain |
|
| nsv1018187 | Chr.5:14385055 - 15126994 on Build GRCh38 | Gain |
|
| nsv4728 | Chr.5:14691303 - 14725902 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map