Assay Details
Target Gene Details
Entrez Gene ID: | 94103 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 3 |
Gene Aliases: |
- |
Location: |
Chr.17:39921041-39927804 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL3 | NM_001320801.1 | 6 | 2357 | NP_001307730.1 |
| NM_001320802.1 | 4 | 1195 | NP_001307731.1 | |
| NM_001320803.1 | 4 | 909 | NP_001307732.1 | |
| NM_139280.3 | 4 | 1200 | NP_644809.1 | |
| XM_017025345.1 | 4 | 1053 | XP_016880834.1 | |
| XM_017025346.1 | 4 | 966 | XP_016880835.1 | |
| XM_017025347.1 | 4 | 1146 | XP_016880836.1 | |
| XM_017025348.1 | 4 | 892 | XP_016880837.1 | |
| AF373101.1 | 4 | 936 | AAN76521.1 | |
| AK074811.1 | 4 | 997 | BAC11223.1 | |
| AK075212.1 | 4 | 952 | BAC11476.1 | |
| AX775843.1 | 4 | 954 | CAE11621.1 | |
| BC017087.1 | 4 | 966 | AAH17087.1 | |
| BC071833.1 | 4 | 945 | AAH71833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map