Assay Details
Target Gene Details
Entrez Gene ID: | 3249 |
Gene Name: | hepsin |
Gene Aliases: |
TMPRSS1 |
Location: |
Chr.19:35040506-35066573 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN | NM_002151.2 | 13 | 1355 | NP_002142.1 |
| NM_182983.2 | 12 | 1935 | NP_892028.1 | |
| XM_005258838.4 | 12 | 1433 | XP_005258895.2 | |
| XM_006723181.3 | 12 | 1309 | XP_006723244.2 | |
| XM_017026731.1 | 12 | 1608 | XP_016882220.1 | |
| XM_017026732.1 | 11 | 1170 | XP_016882221.1 | |
| AK091988.1 | 1 | 1910 | ||
| AK095160.1 | 6 | 1531 | ||
| AK293622.1 | 11 | 1189 | ||
| AK315655.1 | 12 | 1288 | ||
| BC025716.1 | 12 | 1307 | AAH25716.1 | |
| EU644753.1 | 11 | 1110 | ||
| M18930.1 | 13 | 1355 | AAA36013.1 | |
| X07002.1 | 7 | 771 | CAA30058.1 | |
| X07732.1 | 12 | 1935 | CAA30558.1 |
Target Gene Details
Entrez Gene ID: | 100128675 |
Gene Name: | HPN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:35059059-35106304 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN-AS1 | NR_024562.1 | |||
| AK295317.1 |
More Information
Set Membership: |
|

Genomic Map