Assay Details
Target Gene Details
Entrez Gene ID: | 54857 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 2 |
Gene Aliases: |
GDE3, OBDPF |
Location: |
Chr.X:70423031-70433391 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDPD2 | NM_001171191.1 | 14 | 1775 | NP_001164662.1 |
| NM_001171192.1 | 17 | 2146 | NP_001164663.1 | |
| NM_001171193.1 | 15 | 1879 | NP_001164664.1 | |
| NM_017711.3 | 16 | 1993 | NP_060181.2 | |
| XM_011530977.1 | 13 | 1509 | XP_011529279.1 | |
| XM_017029614.1 | 15 | 1974 | XP_016885103.1 | |
| AB048363.1 | 16 | 1883 | BAB13350.1 | |
| AI277430.1 | 1 | 298 | ||
| AK000214.1 | 16 | 1704 | BAA91014.1 | |
| AK097293.1 | 11 | 1471 | ||
| AK299255.1 | ||||
| AK301025.1 | ||||
| AK316035.1 | 15 | 1611 | ||
| AY358986.1 | 16 | 1688 | AAQ89345.1 | |
| BC032009.1 | 16 | 1690 | AAH32009.1 |
Target Gene Details
Entrez Gene ID: | 105373244 |
Gene Name: | uncharacterized LOC105373244 |
Gene Aliases: |
- |
Location: |
Chr.X:70427358-70436800 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373244 | XM_011531081.2 | XP_011529383.1 | ||
| XM_011531082.2 | XP_011529384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map