Assay Details
Target Gene Details
Entrez Gene ID: | 60558 |
Gene Name: | GUF1 homolog, GTPase |
Gene Aliases: |
EF-4 |
Location: |
Chr.4:44678395-44700685 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUF1 | NM_021927.2 | 1 | 165 | NP_068746.2 |
| XM_005248122.3 | 1 | 131 | XP_005248179.1 | |
| XM_011513732.2 | 1 | 186 | XP_011512034.1 | |
| AK023282.1 | 1 | 115 | BAB14507.1 | |
| AK225314.1 | 1 | 120 | ||
| AK308667.1 | 1 | 87 | ||
| BC036768.1 | 1 | 126 | AAH36768.1 | |
| DA997676.1 | 1 | 165 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013488 | Chr.4:44541791 - 44974106 on Build GRCh38 | Loss |
|
| nsv4319 | Chr.4:44671518 - 44716896 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map