Assay Details
Target Gene Details
Entrez Gene ID: | 3609 |
Gene Name: | interleukin enhancer binding factor 3 |
Gene Aliases: |
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80 |
Location: |
Chr.19:10654261-10692419 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
| nsv1160585 | Chr.19:10674243 - 10771056 on Build GRCh38 | Deletion |
|
| nsv833746 | Chr.19:10678751 - 10842756 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map