Assay Details
Target Gene Details
Entrez Gene ID: | 79370 |
Gene Name: | BCL2 like 14 |
Gene Aliases: |
BCLG |
Location: |
Chr.12:12049844-12099695 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L14 | NM_030766.1 | 2 | 439 | NP_110393.1 |
| NM_138722.1 | 2 | 439 | NP_620048.1 | |
| NM_138723.1 | 2 | 445 | NP_620049.1 | |
| XM_006719151.3 | 4 | 669 | XP_006719214.1 | |
| XM_011520848.1 | 4 | 889 | XP_011519150.1 | |
| XM_011520849.1 | 3 | 594 | XP_011519151.1 | |
| XM_017019963.1 | 3 | 414 | XP_016875452.1 | |
| AF281254.1 | 2 | 439 | AAG59793.1 | |
| AF281255.1 | 2 | 439 | AAG59794.1 | |
| AK223415.1 | 2 | 430 | BAD97135.1 | |
| AK292995.1 | 2 | 450 | ||
| AY040274.1 | 1 | 251 | AAK72109.1 | |
| BC025778.1 | 2 | 445 | AAH25778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map