Assay Details
Target Gene Details
Entrez Gene ID: | 4312 |
Gene Name: | matrix metallopeptidase 1 |
Gene Aliases: |
CLG, CLGN |
Location: |
Chr.11:102789910-102798235 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP1 | NM_001145938.1 | 10 | 1384 | NP_001139410.1 |
| NM_002421.3 | 10 | 1536 | NP_002412.1 | |
| AK024818.1 | 10 | 1459 | ||
| AK098450.1 | 11 | 1391 | ||
| AK130687.1 | 10 | 1459 | ||
| AK223034.1 | 9 | 1162 | BAD96754.1 | |
| AK223035.1 | 10 | 1417 | BAD96755.1 | |
| AK223036.1 | 8 | 1000 | BAD96756.1 | |
| AK223056.1 | 9 | 1202 | BAD96776.1 | |
| AK223281.1 | 10 | 1459 | BAD97001.1 | |
| AK225495.1 | 9 | 1227 | ||
| AK297723.1 | 10 | 1307 | ||
| AK301340.1 | 10 | 1249 | ||
| BC013118.1 | 6 | 684 | AAH13118.1 | |
| BC013875.2 | 10 | 1435 | AAH13875.1 | |
| BT006874.1 | AAP35520.1 | |||
| BT020147.1 | AAV38949.1 | |||
| M13509.1 | 10 | 1461 | AAA35699.1 | |
| X05231.1 | 10 | 1459 | CAA28858.1 | |
| X54925.1 | 10 | 1461 | CAA38691.1 |
Target Gene Details
Entrez Gene ID: | 100288077 |
Gene Name: | Wilms tumor 1 associated protein pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.11:102783676-102836766 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WTAPP1 | NR_038390.1 | |||
| AK097805.1 |
More Information
Set Membership: |
|

Genomic Map