Assay Details
Target Gene Details
Entrez Gene ID: | 9801 |
Gene Name: | mitochondrial ribosomal protein L19 |
Gene Aliases: |
L19mt, MRP-L15, MRP-L19, MRPL15, RLX1, RPML15 |
Location: |
Chr.2:75646783-75662208 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL19 | NM_014763.3 | 6 | 3284 | NP_055578.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv706e214 | Chr.2:75581910 - 75690050 on Build GRCh38 | Gain |
|
| dgv20n14 | Chr.2:75596036 - 75687909 on Build GRCh38 | Gain |
|
| esv2759063 | Chr.2:75462279 - 75785697 on Build GRCh38 | Gain+Loss |
|
| nsv438356 | Chr.2:75617501 - 75661034 on Build GRCh38 | Loss |
|
| dgv152e55 | Chr.2:75594376 - 75686819 on Build GRCh38 | Gain |
|
| nsv514988 | Chr.2:75632582 - 75690230 on Build GRCh38 | Loss |
|
| dgv5n43 | Chr.2:75591363 - 75680225 on Build GRCh38 | Gain |
|
| nsv441762 | Chr.2:75594779 - 75687246 on Build GRCh38 | Gain |
|
| nsv834263 | Chr.2:75617574 - 75780540 on Build GRCh38 | Gain |
|
| esv3303063 | Chr.2:75593915 - 75690043 on Build GRCh38 | Tandem Duplication |
|
More Information
Set Membership: |
|

Genomic Map