Assay Details
Target Gene Details
Entrez Gene ID: | 2707 |
Gene Name: | gap junction protein beta 3 |
Gene Aliases: |
CX31, DFNA2, DFNA2B, EKV |
Location: |
Chr.1:34781189-34786366 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB3 | NM_001005752.1 | 2 | 1371 | NP_001005752.1 |
| NM_024009.2 | 2 | 1828 | NP_076872.1 | |
| AF052692.1 | 1 | 2346 | AAD11816.1 | |
| AK026139.1 | 2 | 1827 | ||
| BC012918.1 | 2 | 1360 | AAH12918.1 | |
| BC110640.1 | 1 | 2405 | AAI10641.1 | |
| BE673240.1 | 1 | 393 | ||
| BM846348.1 | 1 | 378 |
More Information
Set Membership: |
|

Genomic Map